Assay Details
| Gene Symbol: Ssh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198109.4 | NP_932777.2 | 13 - 14 | 1376 | 64 | |
| GenBank mRNA | AB099287.1 | - | 13 - 14 | 1411 | 64 | |
| AK155557.1 | - | 13 - 14 | 1401 | 64 | ||
| AK165380.1 | - | 12 - 13 | 1341 | 64 | ||
| AK170787.1 | - | 12 - 13 | 1301 | 64 | ||
| AK171210.1 | - | 13 - 14 | 1425 | 64 | ||
| AK171556.1 | - | 13 - 14 | 1391 | 64 | ||
| AK173144.1 | - | 12 - 13 | 1332 | 64 | ||
| BC046529.1 | - | 7 - 8 | 858 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ssh1
Genomic Map