Assay Details
| Gene Symbol: Cept1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133869.4 | NP_598630.2 | 4 - 5 | 831 | 103 | |
| GenBank mRNA | AK037116.1 | - | 4 - 5 | 791 | 103 | |
| BC023783.1 | - | 4 - 5 | 771 | 5346896 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cept1
Genomic Map