Assay Details
| Gene Symbol: Lpxn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134152.3 | NP_598913.1 | 1 - 2 | 70 | 103 | |
| GenBank mRNA | AB053936.2 | - | 1 - 2 | 122 | 103 | |
| AB071194.1 | - | 1 - 2 | 95 | 103 | ||
| AK039922.1 | - | 1 - 2 | 100 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB053936.2, AB071194.1, NM_134152.3 transcripts.
Set Membership: |
|

Lpxn
Genomic Map