Assay Details
| Gene Symbol: Slc52a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029643.3 | NP_083919.1 | 2 - 3 | 471 | 67 | |
| GenBank mRNA | AK008081.1 | - | 2 - 3 | 450 | 67 | |
| AK029579.1 | - | 2 - 3 | 472 | 67 | ||
| AK032015.1 | - | 2 - 3 | 451 | 67 | ||
| AK080449.1 | - | 2 - 3 | 545 | 67 | ||
| AK142597.1 | - | 2 - 3 | 553 | 67 | ||
| AK152335.1 | - | 2 - 3 | 477 | 67 | ||
| AK161447.1 | - | 2 - 3 | 583 | 67 | ||
| BC016264.1 | - | 2 - 3 | 531 | 4921900 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30349020 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30349001 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30349002 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30349019 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P288828 | BC016264.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P288828 | AK142597.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P288828 | AK152335.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P288828 | AK161447.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P288828 | AK032015.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P288828 | BC016264.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P288828 | AK142597.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P288828 | AK152335.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P288828 | AK161447.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P288828 | AK032015.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 455726 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 140066 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177985 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P288828 | BC016264.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P288828 | AK142597.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P288828 | AK152335.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P288828 | AK161447.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P288828 | AK032015.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P288828 | BC016264.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P288828 | AK142597.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P288828 | AK152335.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P288828 | AK161447.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P288828 | AK032015.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349012 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349014 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349022 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349030 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349029 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349037 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30349021 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177985 | - | - |
| Whole Mouse Genome G4122A | A_51_P288828 | BC016264.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288828 | AK142597.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288828 | AK152335.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288828 | AK161447.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288828 | AK032015.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288828 | BC016264.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288828 | AK142597.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288828 | AK152335.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288828 | AK161447.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P288828 | AK032015.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125737 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177985 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12240208 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12240206 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12115197 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30349002 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30349001 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30349019 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30349020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30349026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12136794 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349050 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12136796 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349048 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12240217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349013 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349012 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12119309 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349032 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12240220 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349018 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349033 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12240213 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349046 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12240206 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349049 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30349044 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10424926 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424928 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424927 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424924 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424925 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10424923 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437381_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451256_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437315_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454870_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456278_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439451_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10424922 | - | - |
| Mouse Genome 430 2.0 Array | 1437315_at | - | - |
| Mouse Genome 430 2.0 Array | 1454870_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1437315_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1439451_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1456278_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1451256_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456278_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1437381_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1454870_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1437381_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1451256_at | - | - |
| Mouse Genome 430 2.0 Array | 1439451_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52710_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52710_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500004277.mm.1 | NM_029643.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004277.mm.1 | NM_029643.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500004277.mm.1 | TC1500000596.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1437381_x_at | - | - |
| Mouse Expression Set 430 | 1439451_PM_x_at | - | - |
| Mouse Expression Set 430 | 1439451_x_at | - | - |
| Mouse Expression Set 430 | 1454870_PM_x_at | - | - |
| Mouse Expression Set 430 | 1454870_x_at | - | - |
| Mouse Expression Set 430 | 1456278_PM_x_at | - | - |
| Mouse Expression Set 430 | 1456278_x_at | - | - |
| Mouse Expression Set 430 | 1437381_PM_x_at | - | - |
| Mouse Expression Set 430 | 1437315_PM_at | - | - |
| Mouse Expression Set 430 | 1437315_at | - | - |
| Murine Genome U74v2 Set | 96201_at | - | - |
| Murine Genome U74v2 Set | 96201_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439451_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456278_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439451_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1454870_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437381_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1454870_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437381_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456278_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1437381_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1454870_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1454870_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451256_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439451_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456278_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1437381_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439451_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456278_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451256_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437315_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437315_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1170136 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1170136 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001170136 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2610956 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2610956 | - | - |
| mouse-6 v1.1 (Illumina) | scl052710.5_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1170136 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl052710.5_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1170136 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2610954 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2957617 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2957617 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2610954 | - | - |

Slc52a2
Genomic Map