Assay Details
Gene Symbol: Rbak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045482.2 | NP_001038947.1 | 2 - 3 | 283 | 63 | |
NM_021326.3 | NP_067301.1 | 2 - 3 | 383 | 63 | ||
GenBank mRNA | AF226870.1 | - | 2 - 3 | 241 | 63 | |
AK051009.1 | - | 2 - 3 | 384 | 63 | ||
CN699599.1 | - | 2 - 3 | 77 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF226870.1, AK051009.1, NM_001045482.2, NM_021326.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
