Assay Details
| Gene Symbol: Lrrc8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033550.2 | NP_001028722.1 | 3 - 4 | 295 | 101 | |
| GenBank mRNA | AK083378.1 | - | 3 - 4 | 296 | 101 | |
| AK220329.1 | - | 1 - 2 | 53 | 101 | ||
| BC138877.1 | - | 2 - 3 | 125 | 8861899 | 101 | |
| BC138878.1 | - | 2 - 3 | 125 | 8861900 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC138877.1, BC138878.1, NM_001033550.2 transcripts.
Set Membership: |
|

Lrrc8b
Genomic Map