Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500031540.mm.1 |
TC0500003461.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500031540.mm.1 |
NM_175521.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031540.mm.1 |
NM_175521.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031543.mm.1 |
TC0500003461.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500031543.mm.1 |
NM_175521.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031543.mm.1 |
NM_175521.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031542.mm.1 |
TC0500003461.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500031542.mm.1 |
NM_175521.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031542.mm.1 |
NM_175521.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031541.mm.1 |
TC0500003461.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500031541.mm.1 |
NM_175521.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500031541.mm.1 |
NM_175521.3 |
Refseq |