Assay Details
| Gene Symbol: Nyap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175521.3 | NP_780730.2 | 2 - 3 | 695 | 68 | |
| GenBank mRNA | AB429289.1 | - | 2 - 3 | 695 | 68 | |
| AK080821.1 | - | 3 - 4 | 1063 | 68 | ||
| AK141276.1 | - | 3 - 4 | 552 | 68 | ||
| BC057372.1 | - | 2 - 3 | 695 | 5686602 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK080821.1 transcripts.
Set Membership: |
|

Nyap1
Genomic Map