Assay Details
Gene Symbol: Gnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010312.4 | NP_034442.1 | 1 - 2 | 92 | 139 | |
GenBank mRNA | AK146941.1 | - | 1 - 2 | 116 | 139 | |
BC059942.1 | - | 1 - 2 | 118 | 6529685 | 139 | |
BC062178.1 | - | 1 - 2 | 75 | 6330944 | 139 | |
Y11970.1 | - | 1 - 2 | 105 | 139 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK146941.1, BC059942.1, BC062178.1, NM_010312.4, Y11970.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
