Assay Details
| Gene Symbol: 1700010H22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005827.1 | - | 498 | 73 | ||
| AK015350.1 | - | 1 - 2 | 74 | |||
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56549 | AK005827.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56549 | AK015350.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56549 | AK005827.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56549 | AK015350.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114319 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172925 | - | - |
| Whole Mouse Genome G4122A | A_52_P56549 | AK005827.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56549 | AK015350.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56549 | AK005827.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56549 | AK015350.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118599 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10531585 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531586 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531584 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531587 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1443839_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454238_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1443840_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10531583 | - | - |
| Mouse Genome 430 2.0 Array | 1443839_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1443840_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1443839_at | - | - |
| Mouse Genome 430 2.0 Array | 1443840_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1454238_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1454238_a_at | - | - |
| Mouse Expression Set 430 | 1443840_x_at | - | - |
| Mouse Expression Set 430 | 1443839_PM_at | - | - |
| Mouse Expression Set 430 | 1454238_PM_a_at | - | - |
| Mouse Expression Set 430 | 1443840_PM_x_at | - | - |
| Mouse Expression Set 430 | 1443839_at | - | - |
| Mouse Expression Set 430 | 1454238_a_at | - | - |
| Murine Genome U74v2 Set | 167803_PM_at | - | - |
| Murine Genome U74v2 Set | 167803_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443839_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443840_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443840_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454238_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454238_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443839_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 493414 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6760309 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101850563 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006760309 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2694984 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2694984 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231945 | - | - |
| mouse-6 v1.1 (Illumina) | scl26342.5.1_143-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101850563 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6760309 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl26342.5.1_143-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6760309 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101850563 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1231945 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2694983 | - | - |

1700010H22Rik
Genomic Map