| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900005512.mm.1 |
NM_001141983.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900005512.mm.1 |
NM_001141983.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900005512.mm.1 |
NM_027694.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900005512.mm.1 |
NM_027694.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900005512.mm.1 |
TC1900000614.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900005514.mm.1 |
NM_001141983.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900005514.mm.1 |
NM_001141983.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900005514.mm.1 |
NM_027694.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900005514.mm.1 |
NM_027694.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900005514.mm.1 |
TC1900000614.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900005507.mm.1 |
NM_001141983.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900005507.mm.1 |
NM_001141983.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900005507.mm.1 |
TC1900000614.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900005509.mm.1 |
NM_027694.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900005509.mm.1 |
NM_027694.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900005509.mm.1 |
TC1900000614.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900005511.mm.1 |
NM_001141983.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900005511.mm.1 |
NM_001141983.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900005511.mm.1 |
NM_027694.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900005511.mm.1 |
NM_027694.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900005511.mm.1 |
TC1900000614.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900005510.mm.1 |
NM_027694.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900005510.mm.1 |
NM_027694.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900005510.mm.1 |
TC1900000614.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900005508.mm.1 |
NM_001141983.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900005508.mm.1 |
NM_001141983.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900005508.mm.1 |
TC1900000614.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900005513.mm.1 |
NM_001141983.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900005513.mm.1 |
NM_001141983.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900005513.mm.1 |
NM_027694.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900005513.mm.1 |
NM_027694.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900005513.mm.1 |
TC1900000614.mm.1 |
MTA TC |