Assay Details
| Gene Symbol: Fam149a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK034229.1 | - | 1 - 2 | 367 | 62 | |
| AK038552.1 | - | 1 - 2 | 355 | 62 | ||
| AK198284.1 | - | 1 - 2 | 81 | 62 | ||
| AK204348.1 | - | 1 - 2 | 62 | |||
| AK212095.1 | - | 1 - 2 | 81 | 62 | ||
| AK220518.1 | - | 1 - 2 | 376 | 62 | ||
More Information
Set Membership: |
|

Fam149a
Genomic Map