Assay Details
| Gene Symbol: Mboat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126314.2 | NP_001119786.1 | 3 | 1258 | 65 | |
| GenBank mRNA | BC147844.1 | - | 3 | 1244 | 9087561 | 65 |
| BC147849.1 | - | 3 | 1243 | 9087566 | 65 | |
| EU518496.1 | - | 3 | 1194 | 65 | ||
| EU721729.1 | - | 3 | 1258 | 65 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
peptidyl-serine octanoylation
dihydrolipoamide branched chain acyltransferase activity
sterol O-acyltransferase activity
N-acetyltransferase activity
O-acyltransferase activity
palmitoleoyl [acyl-carrier-protein]-dependent acyltransferase activity
carnitine O-acyltransferase activity
acetyltransferase activity
C-acyltransferase activity
palmitoyltransferase activity
N-acyltransferase activity
acylglycerol O-acyltransferase activity
serine O-acyltransferase activity
O-acetyltransferase activity
O-octanoyltransferase activity
octanoyltransferase activity
O-palmitoyltransferase activity
S-acyltransferase activity
S-acetyltransferase activity
S-malonyltransferase activity
malonyltransferase activity
C-palmitoyltransferase activity
transferase activity
transferase activity, transferring acyl groups
succinyltransferase activity
N-succinyltransferase activity
O-succinyltransferase activity
S-succinyltransferase activity
sinapoyltransferase activity
O-sinapoyltransferase activity
peptidyl-lysine N6-myristoyltransferase activity
peptidyl-lysine N6-palmitoyltransferase activity
benzoyl acetate-CoA thiolase activity
3-hydroxybutyryl-CoA thiolase activity
3-ketopimelyl-CoA thiolase activity
N-palmitoyltransferase activity
myristoyltransferase activity
acyl-CoA N-acyltransferase activity
protein-cysteine S-myristoyltransferase activity
protein-cysteine S-acyltransferase activity
dihydrolipoamide S-acyltransferase activity
glucosaminyl-phosphotidylinositol O-acyltransferase activity
ergosterol O-acyltransferase activity
lanosterol O-acyltransferase activity
naphthyl-2-oxomethyl-succinyl-CoA succinyl transferase activity
2,4,4-trimethyl-3-oxopentanoyl-CoA 2-C-propanoyl transferase activity
2-methylhexanoyl-CoA C-acetyltransferase activity
butyryl-CoA 2-C-propionyltransferase activity
2,6-dimethyl-5-methylene-3-oxo-heptanoyl-CoA C-acetyltransferase activity
L-2-aminoadipate N-acetyltransferase activity
UDP-3-O-[3-hydroxymyristoyl] glucosamine N-acyltransferase activity
keto acid formate lyase activity
Ras palmitoyltransferase activity
azetidine-2-carboxylic acid acetyltransferase activity
peptidyl-lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
acetyl-CoA:L-lysine N6-acetyltransferase
peptidyl-serine octanoylation
dihydrolipoamide branched chain acyltransferase activity
sterol O-acyltransferase activity
N-acetyltransferase activity
O-acyltransferase activity
palmitoleoyl [acyl-carrier-protein]-dependent acyltransferase activity
carnitine O-acyltransferase activity
acetyltransferase activity
C-acyltransferase activity
palmitoyltransferase activity
N-acyltransferase activity
acylglycerol O-acyltransferase activity
serine O-acyltransferase activity
O-acetyltransferase activity
O-octanoyltransferase activity
octanoyltransferase activity
O-palmitoyltransferase activity
S-acyltransferase activity
S-acetyltransferase activity
S-malonyltransferase activity
malonyltransferase activity
C-palmitoyltransferase activity
transferase activity
transferase activity, transferring acyl groups
succinyltransferase activity
N-succinyltransferase activity
O-succinyltransferase activity
S-succinyltransferase activity
sinapoyltransferase activity
O-sinapoyltransferase activity
peptidyl-lysine N6-myristoyltransferase activity
peptidyl-lysine N6-palmitoyltransferase activity
benzoyl acetate-CoA thiolase activity
3-hydroxybutyryl-CoA thiolase activity
3-ketopimelyl-CoA thiolase activity
N-palmitoyltransferase activity
myristoyltransferase activity
acyl-CoA N-acyltransferase activity
protein-cysteine S-myristoyltransferase activity
protein-cysteine S-acyltransferase activity
dihydrolipoamide S-acyltransferase activity
glucosaminyl-phosphotidylinositol O-acyltransferase activity
ergosterol O-acyltransferase activity
lanosterol O-acyltransferase activity
naphthyl-2-oxomethyl-succinyl-CoA succinyl transferase activity
2,4,4-trimethyl-3-oxopentanoyl-CoA 2-C-propanoyl transferase activity
2-methylhexanoyl-CoA C-acetyltransferase activity
butyryl-CoA 2-C-propionyltransferase activity
2,6-dimethyl-5-methylene-3-oxo-heptanoyl-CoA C-acetyltransferase activity
L-2-aminoadipate N-acetyltransferase activity
UDP-3-O-[3-hydroxymyristoyl] glucosamine N-acyltransferase activity
keto acid formate lyase activity
Ras palmitoyltransferase activity
azetidine-2-carboxylic acid acetyltransferase activity
peptidyl-lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
acetyl-CoA:L-lysine N6-acetyltransferase
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2126167 | EU721729.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2126167 | BC147849.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2126167 | NM_001126314.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2126167 | BC147844.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2126167 | EU518496.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 571819 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 771169 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 604717 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 274935 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 792472 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2126167 | EU721729.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2126167 | BC147849.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2126167 | NM_001126314.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2126167 | BC147844.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2126167 | EU518496.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25772275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772248 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772269 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07006675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07006829 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06915757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06915756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06857949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07153288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07210707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06715836 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772253 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06794630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07379314 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06794627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07153287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07220576 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772250 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25772265 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10571301 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439752_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10571300 | - | - |
| Mouse Genome 430 2.0 Array | 1439752_at | - | - |
| Mouse Genome 430 2.0 Array | 1439752_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800002986.mm.1 | NM_001126314.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800002986.mm.1 | NM_001126314.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800002986.mm.1 | TC0800000402.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1439752_PM_at | - | - |
| Mouse Expression Set 430 | 1439752_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439752_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439752_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 344342 | - | - |

Mboat4
Genomic Map