Assay Details
| Gene Symbol: Fth1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010239.2 | NP_034369.1 | 3 - 4 | 617 | 100 | |
| NR_073181.1 | 4 - 5 | 1186 | 100 | |||
| GenBank mRNA | AK027998.1 | - | 3 - 4 | 560 | 100 | |
| AK088935.1 | - | 2 - 3 | 1765 | 100 | ||
| AK139622.1 | - | 3 - 4 | 560 | 100 | ||
| AK147082.1 | - | 3 - 4 | 563 | 100 | ||
| AK150262.1 | - | 3 - 4 | 560 | 100 | ||
| AK150508.1 | - | 3 - 4 | 560 | 100 | ||
| AK150628.1 | - | 3 - 4 | 560 | 100 | ||
| AK150679.1 | - | 3 - 4 | 563 | 100 | ||
| AK150693.1 | - | 3 - 4 | 560 | 100 | ||
| AK151192.1 | - | 3 - 4 | 560 | 100 | ||
| AK151241.1 | - | 3 - 4 | 563 | 100 | ||
| AK151399.1 | - | 3 - 4 | 546 | 100 | ||
| AK151609.1 | - | 3 - 4 | 560 | 100 | ||
| AK151675.1 | - | 3 - 4 | 560 | 100 | ||
| AK152071.1 | - | 3 - 4 | 560 | 100 | ||
| AK152702.1 | - | 4 - 5 | 1188 | 100 | ||
| AK153017.1 | - | 3 - 4 | 560 | 100 | ||
| AK153195.1 | - | 3 - 4 | 563 | 100 | ||
| AK153199.1 | - | 3 - 4 | 563 | 100 | ||
| AK159243.1 | - | 3 - 4 | 560 | 100 | ||
| AK168601.1 | - | 3 - 4 | 560 | 100 | ||
| AK169004.1 | - | 3 - 4 | 560 | 100 | ||
| BC011096.1 | - | 3 - 4 | 543 | 100 | ||
| BC012314.1 | - | 3 - 4 | 538 | 3488821 | 100 | |
| J03941.1 | - | 3 - 4 | 513 | 100 | ||
| M24509.1 | - | 3 - 4 | 496 | 100 | ||
| X12812.1 | - | 3 - 4 | 459 | 100 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fth1
Genomic Map