Assay Details
| Gene Symbol: Cd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037801.2 | NP_001032890.1 | 3 - 4 | 665 | 91 | |
| NM_009852.3 | NP_033982.3 | 3 - 4 | 665 | 91 | ||
| GenBank mRNA | AK030822.1 | - | 3 - 4 | 635 | 91 | |
| AK030823.1 | - | 3 - 4 | 666 | 91 | ||
| AK088050.1 | - | 3 - 4 | 643 | 91 | ||
| BC014294.1 | - | 3 - 4 | 671 | 4187689 | 91 | |
| U12434.1 | - | 3 - 4 | 463 | 91 | ||
| U35370.1 | - | 3 - 4 | 581 | 91 | ||
| U37543.1 | - | 3 - 4 | 622 | 91 | ||
| U37544.1 | - | 3 - 4 | 622 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd6
Genomic Map