Assay Details
| Gene Symbol: Jak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048177.2 | NP_001041642.1 | 19 - 20 | 3098 | 163 | |
| NM_008413.3 | NP_032439.2 | 20 - 21 | 3181 | 163 | ||
| GenBank mRNA | BC054807.1 | - | 19 - 20 | 2861 | 6838318 | 163 |
| BC059834.1 | - | 19 - 20 | 2861 | 6838318 | 163 | |
| L16956.1 | - | 19 - 20 | 2850 | 163 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Jak2
Genomic Map