Assay Details
| Gene Symbol: Ube2r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026275.4 | NP_080551.1 | 2 - 3 | 310 | 85 | |
| GenBank mRNA | AJ240086.1 | - | 2 - 3 | 269 | 85 | |
| AK003550.1 | - | 2 - 3 | 330 | 85 | ||
| AK007517.1 | - | 2 - 3 | 444 | 85 | ||
| AK075703.1 | - | 2 - 3 | 320 | 85 | ||
| AK075714.1 | - | 2 - 3 | 311 | 85 | ||
| BC011112.1 | - | 2 - 3 | 318 | 3982808 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ube2r2
Genomic Map