Assay Details
| Gene Symbol: Bag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171739.1 | NP_001165210.1 | 3 - 4 | 789 | 116 | |
| NM_009736.3 | NP_033866.4 | 3 - 4 | 789 | 116 | ||
| GenBank mRNA | AF022223.1 | - | 3 - 4 | 730 | 116 | |
| AK009149.1 | - | 3 - 4 | 700 | 116 | ||
| AK013611.1 | - | 1 - 2 | 75 | 116 | ||
| AK154469.1 | - | 3 - 4 | 739 | 116 | ||
| AK171094.1 | - | 3 - 4 | 739 | 116 | ||
| BC003722.1 | - | 3 - 4 | 751 | 3495306 | 116 | |
| BC069918.1 | - | 3 - 4 | 726 | 4979705 | 116 | |
| BC093509.1 | - | 3 - 4 | 700 | 4978330 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bag1
Genomic Map