Assay Details
| Gene Symbol: Bag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171739.1 | NP_001165210.1 | 5 - 6 | 1009 | 155 | |
| NM_009736.3 | NP_033866.4 | 5 - 6 | 1009 | 155 | ||
| GenBank mRNA | AF022223.1 | - | 5 - 6 | 950 | 155 | |
| AK009149.1 | - | 5 - 6 | 920 | 155 | ||
| AK013611.1 | - | 3 - 4 | 295 | 155 | ||
| AK154469.1 | - | 5 - 6 | 959 | 154 | ||
| AK171094.1 | - | 5 - 6 | 959 | 154 | ||
| BC003722.1 | - | 5 - 6 | 971 | 3495306 | 155 | |
| BC069918.1 | - | 5 - 6 | 946 | 4979705 | 155 | |
| BC093509.1 | - | 5 - 6 | 920 | 4978330 | 155 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bag1
Genomic Map