Assay Details
| Gene Symbol: Mat1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133653.3 | NP_598414.1 | 1 - 2 | 231 | 97 | |
| GenBank mRNA | BB871001.1 | - | 1 - 2 | 53 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_133653.3 transcripts.
Set Membership: |
|

Mat1a
Genomic Map