Assay Details
| Gene Symbol: Mat1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133653.3 | NP_598414.1 | 4 - 5 | 764 | 73 | |
| GenBank mRNA | AK149542.1 | - | 3 - 4 | 515 | 73 | |
| AK153832.1 | - | 3 - 4 | 518 | 73 | ||
| BC011211.1 | - | 3 - 4 | 502 | 4196928 | 73 | |
| L13622.1 | - | 3 - 4 | 516 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mat1a
Genomic Map