Assay Details
| Gene Symbol: Mat1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133653.3 | NP_598414.1 | 5 - 6 | 875 | 86 | |
| GenBank mRNA | AK149542.1 | - | 4 - 5 | 626 | 86 | |
| AK153832.1 | - | 4 - 5 | 629 | 86 | ||
| BC011211.1 | - | 4 - 5 | 613 | 4196928 | 86 | |
| L13622.1 | - | 4 - 5 | 627 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mat1a
Genomic Map