Assay Details
| Gene Symbol: Mat1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133653.3 | NP_598414.1 | 7 - 8 | 1250 | 80 | |
| GenBank mRNA | AK149542.1 | - | 6 - 7 | 1001 | 80 | |
| AK153832.1 | - | 6 - 7 | 1004 | 80 | ||
| BC011211.1 | - | 6 - 7 | 988 | 4196928 | 80 | |
| L13622.1 | - | 6 - 7 | 1002 | 80 | ||
More Information
Set Membership: |
|

Mat1a
Genomic Map