Assay Details
| Gene Symbol: Mmrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153127.3 | NP_694767.3 | 4 - 5 | 591 | 111 | |
| GenBank mRNA | AK077884.1 | - | 4 - 5 | 593 | 111 | |
| AK078470.1 | - | 4 - 5 | 567 | 111 | ||
| AK134285.1 | - | 4 - 5 | 631 | 110 | ||
| BC145845.1 | - | 4 - 5 | 511 | 40131148 | 111 | |
More Information
Set Membership: |
|

Mmrn2
Genomic Map