Assay Details
| Gene Symbol: Hgsnat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029884.1 | NP_084160.1 | 15 - 16 | 1643 | 102 | |
| GenBank mRNA | AK035264.1 | - | 15 - 16 | 1576 | 102 | |
| AK149883.1 | - | 15 - 16 | 1647 | 102 | ||
| AK152926.1 | - | 15 - 16 | 1608 | 102 | ||
| AK159649.1 | - | 15 - 16 | 1572 | 102 | ||
| AK160068.1 | - | 15 - 16 | 1566 | 102 | ||
| BC024084.1 | - | 15 - 16 | 1550 | 5101064 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hgsnat
Genomic Map