Assay Details
| Gene Symbol: Dyrk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113389.1 | NP_001106860.1 | 8 - 9 | 1238 | 75 | |
| NM_007890.2 | NP_031916.1 | 8 - 9 | 1497 | 75 | ||
| GenBank mRNA | AK047845.1 | - | 3 - 4 | 485 | 75 | |
| AK052390.1 | - | 2 - 3 | 373 | 75 | ||
| BC034550.1 | - | 3 - 4 | 583 | 75 | ||
| BC129888.1 | - | 7 - 8 | 1100 | 40111469 | 75 | |
| BC129889.1 | - | 7 - 8 | 1100 | 40111470 | 75 | |
| BC138716.1 | - | 7 - 8 | 1127 | 8861738 | 75 | |
| BC145405.1 | - | 7 - 8 | 1100 | 9053963 | 75 | |
| U58497.1 | - | 7 - 8 | 1127 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dyrk1a
Genomic Map