| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800014023.mm.1 |
NM_001024504.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014023.mm.1 |
NM_001024504.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014023.mm.1 |
NM_001042649.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014023.mm.1 |
NM_001042649.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014023.mm.1 |
NM_001042650.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014023.mm.1 |
NM_001042650.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014023.mm.1 |
NM_001042651.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014023.mm.1 |
NM_001042651.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014023.mm.1 |
TC0800001733.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800014025.mm.1 |
NM_001024504.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014025.mm.1 |
NM_001024504.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014025.mm.1 |
NM_001042649.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014025.mm.1 |
NM_001042649.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014025.mm.1 |
NM_001042650.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014025.mm.1 |
NM_001042650.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014025.mm.1 |
NM_001042651.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014025.mm.1 |
NM_001042651.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014025.mm.1 |
TC0800001733.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800014027.mm.1 |
NM_001024504.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014027.mm.1 |
NM_001024504.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014027.mm.1 |
NM_001042649.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014027.mm.1 |
NM_001042649.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014027.mm.1 |
NM_001042650.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014027.mm.1 |
NM_001042650.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014027.mm.1 |
NM_001042651.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014027.mm.1 |
NM_001042651.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800014027.mm.1 |
TC0800001733.mm.1 |
MTA TC |