Assay Details
| Gene Symbol: Dcun1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024504.2 | NP_001019675.2 | 5 - 6 | 215 | ||
| NM_001042651.1 | NP_001036116.1 | 5 - 6 | 919 | 118 | ||
| GenBank mRNA | AK155726.1 | - | 5 - 6 | 215 | ||
| AK161694.1 | - | 5 - 6 | 624 | 118 | ||
| BC030839.1 | - | 5 - 6 | 215 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Dcun1d2
Genomic Map