Assay Details
| Gene Symbol: Osbpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144500.3 | NP_653083.2 | 1 - 2 | 105 | 148 | |
| GenBank mRNA | AK145292.1 | - | 1 - 2 | 148 | ||
| AK170669.1 | - | 1 - 2 | 106 | 148 | ||
| BC026804.1 | - | 1 - 2 | 112 | 4159217 | 148 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK145292.1, AK170669.1, BC026804.1, NM_144500.3 transcripts.
Set Membership: |
|

Osbpl2
Genomic Map