Assay Details
| Gene Symbol: Osbpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144500.3 | NP_653083.2 | 6 - 7 | 691 | 104 | |
| GenBank mRNA | AK048537.1 | - | 6 - 7 | 611 | 104 | |
| AK145292.1 | - | 6 - 7 | 723 | 104 | ||
| AK161695.1 | - | 6 - 7 | 630 | 104 | ||
| AK166059.1 | - | 6 - 7 | 670 | 104 | ||
| AK170306.1 | - | 6 - 7 | 675 | 104 | ||
| AK170669.1 | - | 6 - 7 | 692 | 104 | ||
| AK185626.1 | - | 1 - 2 | 81 | 104 | ||
| BC026804.1 | - | 6 - 7 | 698 | 4159217 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Osbpl2
Genomic Map