Assay Details
| Gene Symbol: Tapt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173764.3 | NP_776125.2 | 4 - 5 | 611 | 107 | |
| GenBank mRNA | BC096038.1 | - | 1 - 2 | 90 | 107 | |
| BC110670.1 | - | 3 - 4 | 332 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tapt1
Genomic Map