Assay Details
| Gene Symbol: Tapt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173764.3 | NP_776125.2 | 6 - 7 | 829 | 144 | |
| GenBank mRNA | AK030002.1 | - | 1 - 2 | 71 | 144 | |
| BC096038.1 | - | 3 - 4 | 308 | 144 | ||
| BC110670.1 | - | 5 - 6 | 550 | 144 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tapt1
Genomic Map