Assay Details
| Gene Symbol: Tapt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173764.3 | NP_776125.2 | 10 - 11 | 1160 | 67 | |
| GenBank mRNA | AK030002.1 | - | 5 - 6 | 402 | 67 | |
| BC096038.1 | - | 7 - 8 | 639 | 67 | ||
| BC110670.1 | - | 9 - 10 | 881 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tapt1
Genomic Map