Assay Details
| Gene Symbol: Pgm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025700.2 | NP_079976.1 | 6 - 7 | 774 | 82 | |
| GenBank mRNA | AK014332.1 | - | 6 - 7 | 699 | 82 | |
| AK181291.1 | - | 3 - 4 | 295 | 82 | ||
| BC030869.1 | - | 6 - 7 | 736 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pgm1
Genomic Map