Assay Details
| Gene Symbol: Tbc1d12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145952.3 | NP_666064.3 | 4 - 5 | 1193 | 98 | |
| GenBank mRNA | AK172979.1 | - | 4 - 5 | 1187 | 98 | |
| BC138056.1 | - | 4 - 5 | 1089 | 8861075 | 98 | |
| BC138057.1 | - | 4 - 5 | 1089 | 8861076 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbc1d12
Genomic Map