Assay Details
| Gene Symbol: Tbc1d12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145952.3 | NP_666064.3 | 7 - 8 | 1491 | 70 | |
| GenBank mRNA | AK172979.1 | - | 7 - 8 | 1485 | 70 | |
| BC138056.1 | - | 7 - 8 | 1387 | 8861075 | 70 | |
| BC138057.1 | - | 7 - 8 | 1387 | 8861076 | 70 | |
| BC145129.1 | - | 5 - 6 | 1162 | 9053677 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbc1d12
Genomic Map