Assay Details
| Gene Symbol: Thbs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013691.2 | NP_038719.2 | 10 - 11 | 1236 | 61 | |
| GenBank mRNA | BC053023.1 | - | 10 - 11 | 1190 | 6401682 | 61 |
| BC060104.1 | - | 9 - 10 | 1986 | 61 | ||
| L04302.1 | - | 10 - 11 | 1216 | 61 | ||
| L24434.1 | - | 10 - 11 | 1213 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Thbs3
Genomic Map