Assay Details
| Gene Symbol: Thbs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013691.2 | NP_038719.2 | 2 - 3 | 348 | 56 | |
| GenBank mRNA | BC053023.1 | - | 2 - 3 | 302 | 6401682 | 56 |
| CJ064850.1 | - | 2 - 3 | 349 | 56 | ||
| L04302.1 | - | 2 - 3 | 328 | 56 | ||
| L24434.1 | - | 2 - 3 | 325 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Thbs3
Genomic Map