Assay Details
| Gene Symbol: Thbs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013691.2 | NP_038719.2 | 7 - 8 | 863 | 89 | |
| GenBank mRNA | BC053023.1 | - | 7 - 8 | 817 | 6401682 | 89 |
| L24434.1 | - | 7 - 8 | 840 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Thbs3
Genomic Map