Assay Details
| Gene Symbol: Cd209e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK040917.1 | - | 1 - 2 | 78 | 112 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK040917.1 transcripts.
Set Membership: |
|

Cd209e
Genomic Map