Assay Details
| Gene Symbol: Cd209e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130905.2 | NP_570975.1 | 4 - 5 | 391 | 87 | |
| GenBank mRNA | AF373412.1 | - | 4 - 5 | 391 | 87 | |
| AK040917.1 | - | 4 - 5 | 413 | 87 | ||
| BC107188.2 | - | 4 - 5 | 391 | 40050810 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cd209e
Genomic Map