Assay Details
| Gene Symbol: Star | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011485.4 | NP_035615.2 | 4 - 5 | 521 | 65 | |
| GenBank mRNA | AK019725.2 | - | 4 - 5 | 561 | 65 | |
| AK054470.1 | - | 4 - 5 | 521 | 65 | ||
| AK136327.1 | - | 4 - 5 | 531 | 65 | ||
| BC082283.1 | - | 4 - 5 | 532 | 30436512 | 65 | |
| L36062.1 | - | 4 - 5 | 537 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Star
Genomic Map