Assay Details
| Gene Symbol: Ash2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080793.2 | NP_001074262.1 | 14 - 15 | 1643 | 71 | |
| NM_001286207.1 | NP_001273136.1 | 14 - 15 | 1616 | 71 | ||
| NM_011791.3 | NP_035921.2 | 14 - 15 | 1733 | 71 | ||
| GenBank mRNA | AB020983.2 | - | 14 - 15 | 1715 | 71 | |
| AK087934.1 | - | 14 - 15 | 1708 | 71 | ||
| AK145576.1 | - | 14 - 15 | 1608 | 71 | ||
| AK145790.1 | - | 14 - 15 | 1620 | 71 | ||
| AK145967.1 | - | 14 - 15 | 1587 | 71 | ||
| AK146599.1 | - | 14 - 15 | 1617 | 71 | ||
| AK146756.1 | - | 14 - 15 | 1608 | 71 | ||
| AK146938.1 | - | 14 - 15 | 1710 | 71 | ||
| AK212085.1 | - | 3 - 4 | 211 | 71 | ||
| BC012957.1 | - | 14 - 15 | 1720 | 4211433 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ash2l
Genomic Map