Assay Details
| Gene Symbol: Fam189b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014995.1 | NP_001014995.1 | 10 - 11 | 2087 | 74 | |
| GenBank mRNA | BC068123.1 | - | 9 - 10 | 2005 | 74 | |
| BC085505.1 | - | 10 - 11 | 1828 | 74 | ||
| BC088990.1 | - | 10 - 11 | 2087 | 6402466 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam189b
Genomic Map