Assay Details
| Gene Symbol: Fam189b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014995.1 | NP_001014995.1 | 11 - 12 | 2339 | 88 | |
| GenBank mRNA | BC068123.1 | - | 10 - 11 | 2257 | 88 | |
| BC085505.1 | - | 11 - 12 | 2080 | 88 | ||
| BC088990.1 | - | 11 - 12 | 2339 | 6402466 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22317744 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22317743 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22317742 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2007389 | BC085505.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2007389 | BC088990.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2007389 | NM_001014995.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2007389 | AK003667.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2007389 | BC068123.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 185759 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 205521 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 110941 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 386693 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 951121 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 3537 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127463 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2007389 | BC085505.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2007389 | BC088990.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2007389 | NM_001014995.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2007389 | AK003667.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2007389 | BC068123.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317719 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317731 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317718 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317730 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317713 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317725 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317726 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317722 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317711 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317727 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317724 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317735 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317715 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317717 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22317716 | - | - |
| Whole Mouse Genome G4122A | A_51_P372522 | BC085505.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P372522 | BC088990.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P372522 | NM_001014995.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P372522 | AK003667.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P372522 | BC068123.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P77695 | BC085505.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P77695 | BC088990.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P77695 | NM_001014995.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P77695 | BC068123.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03175230 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03175226 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03175215 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22317743 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22317742 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22317744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22317754 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02586683 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317713 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02889607 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03175252 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317754 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02644591 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02644576 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317731 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317715 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02644573 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317755 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317734 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317725 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02642027 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02644597 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317723 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317726 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317730 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317746 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317732 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317747 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317729 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03175245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02644577 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317719 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02644579 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317744 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03175232 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22317759 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02644580 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03175246 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10493424 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493428 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493427 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493426 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493422 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493432 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493430 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493433 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493425 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493434 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493431 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493423 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493429 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454893_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10493421 | - | - |
| Mouse Genome 430 2.0 Array | 1454893_at | - | - |
| Mouse Genome 430 2.0 Array | 1454893_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68521_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68521_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300005595.mm.1 | NM_001014995.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300005595.mm.1 | NM_001014995.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300005595.mm.1 | TC0300000749.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300005596.mm.1 | NM_001014995.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300005596.mm.1 | NM_001014995.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300005596.mm.1 | TC0300000749.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1454893_at | - | - |
| Mouse Expression Set 430 | 1454893_PM_at | - | - |
| Murine Genome U74v2 Set | 94958_PM_at | - | - |
| Murine Genome U74v2 Set | 94958_at | - | - |
| Mouse Genome 430A 2.0 Array | 1454893_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1454893_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1454893_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1454893_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 791691 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1217406 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1217406 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223229 | - | - |

Fam189b
Genomic Map