Assay Details
| Gene Symbol: 9230112D13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030062.1 | NP_084338.1 | 1 - 2 | 153 | 67 | |
| GenBank mRNA | AK020336.1 | - | 1 - 2 | 154 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
bone morphogenetic protein receptor, type 1A |
Bmpr1a |
1110037I22Rik, ALK3, AU045487, BMPR-IA, Bmpr |
-10.8kb | Sense | Chr. 14 - 34411061 - 34503532 on NCBI Build GRCm38 |
| Downstream | LIM domain binding 3 | Ldb3 |
AW742271, PDLIM6, ZASP |
3.9kb | Sense | Chr. 14 - 34526699 - 34588698 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

9230112D13Rik
Genomic Map