Assay Details
| Gene Symbol: Clic6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172469.3 | NP_766057.1 | 4 - 5 | 1745 | 101 | |
| GenBank mRNA | AF448440.1 | - | 4 - 5 | 1438 | 101 | |
| AK077599.1 | - | 4 - 5 | 1746 | 101 | ||
| BC075706.1 | - | 4 - 5 | 1770 | 2648653 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clic6
Genomic Map