Assay Details
Gene Symbol: Lrpap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013587.2 | NP_038615.2 | 3 - 4 | 488 | 89 | |
GenBank mRNA | AJ639614.1 | - | 3 - 4 | 495 | 89 | |
AK089250.1 | - | 3 - 4 | 491 | 89 | ||
AK145710.1 | - | 3 - 4 | 531 | 89 | ||
AK150451.1 | - | 3 - 4 | 502 | 89 | ||
AK152485.1 | - | 3 - 4 | 502 | 89 | ||
AK153323.1 | - | 3 - 4 | 502 | 89 | ||
AK166616.1 | - | 3 - 4 | 491 | 89 | ||
AK166746.1 | - | 3 - 4 | 502 | 89 | ||
AK166819.1 | - | 3 - 4 | 506 | 89 | ||
AK168421.1 | - | 3 - 4 | 491 | 89 | ||
AK169132.1 | - | 3 - 4 | 496 | 89 | ||
AK171471.1 | - | 3 - 4 | 491 | 89 | ||
BC032170.1 | - | 3 - 4 | 491 | 89 | ||
BC046641.1 | - | 3 - 4 | 480 | 89 | ||
BC057979.1 | - | 3 - 4 | 459 | 89 | ||
BC059887.1 | - | 3 - 4 | 305 | 89 | ||
BC094324.1 | - | 3 - 4 | 488 | 6402163 | 89 | |
D00622.1 | - | 3 - 4 | 481 | 89 | ||
S67967.1 | - | 3 - 4 | 514 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
