Assay Details
| Gene Symbol: Osgin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145950.4 | NP_666062.3 | 2 - 3 | 351 | 103 | |
| GenBank mRNA | AK053737.1 | - | 2 - 3 | 295 | 103 | |
| AK148243.1 | - | 2 - 3 | 326 | 103 | ||
| AK169678.1 | - | 2 - 3 | 296 | 103 | ||
| AK186777.1 | - | 1 - 2 | 53 | 103 | ||
| CB574822.1 | - | 2 - 3 | 307 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Osgin2
Genomic Map