Assay Details
| Gene Symbol: Osgin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145950.4 | NP_666062.3 | 3 - 4 | 475 | 121 | |
| GenBank mRNA | AK048731.1 | - | 3 - 4 | 476 | 121 | |
| AK053737.1 | - | 3 - 4 | 419 | 121 | ||
| AK148243.1 | - | 3 - 4 | 450 | 121 | ||
| AK169678.1 | - | 3 - 4 | 420 | 121 | ||
| AK186777.1 | - | 2 - 3 | 177 | 121 | ||
| CB574822.1 | - | 3 - 4 | 431 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Osgin2
Genomic Map